Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB4B
Entrez Gene ID
403956
Gene Name
RAB4B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.1: 113003317 - 113011978 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003275.3 NP_001003275.1 1 - 2 121 59
GenBank mRNA X56389.1 - - 116 59

View Full Product Details