Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UACA
Entrez Gene ID
403704
Gene Name
uveal autoantigen with coiled-coil domains and ankyrin repeats
Gene Aliases
C3VS
Chromosome Location
Chr.30: 34376072 - 34467278 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003112.1 NP_001003112.1 7 - 8 932 72
XM_005638170.3 XP_005638227.1 7 - 8 967 72
XM_005638171.3 XP_005638228.1 7 - 8 677 72
GenBank mRNA X99145.2 - - 932 72

View Full Product Details