Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM11
Entrez Gene ID
479532
Gene Name
transmembrane protein 11
Gene Aliases
-
Chromosome Location
Chr.5: 42669811 - 42684691 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_859742.5 XP_864835.1 1 - 2 57 112

View Full Product Details