Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPL27
Entrez Gene ID
491078
Gene Name
mitochondrial ribosomal protein L27
Gene Aliases
-
Chromosome Location
Chr.9: 26336595 - 26342512 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005624638.3 XP_005624695.2 1 - 1 245 68

View Full Product Details