Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL15
Entrez Gene ID
476897
Gene Name
methyltransferase like 15
Gene Aliases
METT5D1
Chromosome Location
Chr.21: 48578735 - 48893104 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014122786.2 XP_013978261.1 5 - 6 1022 80
XM_534100.6 XP_534100.3 5 - 6 1299 80

View Full Product Details