Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS20
Entrez Gene ID
477887
Gene Name
ribosomal protein S20
Gene Aliases
-
Chromosome Location
Chr.29: 7488481 - 7489850 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_535079.5 XP_535079.2 1 - 2 161 64

View Full Product Details