Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYSMD2
Entrez Gene ID
478308
Gene Name
LysM domain containing 2
Gene Aliases
-
Chromosome Location
Chr.30: 17409338 - 17424092 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005638413.3 XP_005638470.1 2 - 3 771 74
XM_005638414.3 XP_005638471.1 2 - 3 498 74
XM_535483.6 XP_535483.3 2 - 3 468 74

View Full Product Details