Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABHD10
Entrez Gene ID
478561
Gene Name
abhydrolase domain containing 10, depalmitoylase
Gene Aliases
-
Chromosome Location
Chr.33: 16618085 - 16631084 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014110202.2 XP_013965677.2 1 - 2 212 86
XM_535737.6 XP_535737.5 1 - 2 202 86

View Full Product Details