Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRIF1
Entrez Gene ID
479906
Gene Name
ligand dependent nuclear receptor interacting factor 1
Gene Aliases
-
Chromosome Location
Chr.6: 41023383 - 41041776 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005621791.3 XP_005621848.1 3 - 4 2233 130
XM_022420340.1 XP_022276048.1 3 - 4 2053 130
XM_860702.5 XP_865795.1 2 - 3 550 130

View Full Product Details