Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC25A4
Entrez Gene ID
475630
Gene Name
solute carrier family 25 member 4
Gene Aliases
-
Chromosome Location
Chr.16: 45454660 - 45458606 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_532844.6 XP_532844.2 3 - 4 828 83

View Full Product Details