Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COQ10A
Entrez Gene ID
474402
Gene Name
coenzyme Q10A
Gene Aliases
-
Chromosome Location
Chr.10: 580022 - 583264 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_531633.5 XP_531633.2 - 629 69

View Full Product Details