Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC42EP4
Entrez Gene ID
475907
Gene Name
CDC42 effector protein 4
Gene Aliases
-
Chromosome Location
Chr.9: 6888183 - 6909986 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_533114.6 XP_533114.2 - 157 143

View Full Product Details