Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD39
Entrez Gene ID
474568
Gene Name
ankyrin repeat domain 39
Gene Aliases
-
Chromosome Location
Chr.10: 45117875 - 45126556 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_531796.6 XP_531796.1 1 - 2 142 69

View Full Product Details