Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKRD39
Entrez Gene ID
474568
Gene Name
ankyrin repeat domain 39
Gene Aliases
-
Chromosome Location
Chr.10: 45117875 - 45126556 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_531796.6 XP_531796.1 2 - 3 254 81

View Full Product Details