Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KHK
Entrez Gene ID
100170647
Gene Name
ketohexokinase
Gene Aliases
-
Chromosome Location
Chr.17: 21079907 - 21092510 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195155.1 NP_001182084.1 1 - 2 238 76
GenBank mRNA DN444597.1 - 1 - 2 172 76

View Full Product Details