Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RIBC2
Entrez Gene ID
474469
Gene Name
RIB43A domain with coiled-coils 2
Gene Aliases
-
Chromosome Location
Chr.10: 20625423 - 20640266 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_531699.6 XP_531699.4 5 - 6 899 57

View Full Product Details