Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ICAM2
Entrez Gene ID
480475
Gene Name
intercellular adhesion molecule 2
Gene Aliases
-
Chromosome Location
Chr.9: 11903593 - 11908835 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022423905.1 XP_022279613.1 - 243 68

View Full Product Details