Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EGLN3
Entrez Gene ID
480286
Gene Name
egl-9 family hypoxia inducible factor 3
Gene Aliases
-
Chromosome Location
Chr.8: 12873755 - 12901728 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_537408.5 XP_537408.3 2 - 3 747 106

View Full Product Details