Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NEDD8
Entrez Gene ID
480265
Gene Name
NEDD8 ubiquitin like modifier
Gene Aliases
-
Chromosome Location
Chr.8: 4202621 - 4213090 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_537389.6 XP_537389.2 1 - 2 160 80

View Full Product Details