Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SH3BP1
Entrez Gene ID
481265
Gene Name
SH3 domain binding protein 1
Gene Aliases
-
Chromosome Location
Chr.10: 26953993 - 26966417 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022424466.1 XP_022280174.1 11 - 12 1182 73
XM_538386.6 XP_538386.3 11 - 12 1044 73

View Full Product Details