Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLAAT4
Entrez Gene ID
483772
Gene Name
phospholipase A and acyltransferase 4
Gene Aliases
RARRES3
Chromosome Location
Chr.18: 53418184 - 53424027 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014121355.2 XP_013976830.1 2 - 3 262 81

View Full Product Details