Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSPB6
Entrez Gene ID
484574
Gene Name
heat shock protein family B (small) member 6
Gene Aliases
-
Chromosome Location
Chr.1: 116878370 - 116880852 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_541688.6 XP_541688.1 1 - 2 249 88

View Full Product Details