Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CABP5
Entrez Gene ID
484414
Gene Name
calcium binding protein 5
Gene Aliases
-
Chromosome Location
Chr.1: 108147179 - 108157063 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005616376.3 XP_005616433.1 1 - 2 182 65
XM_541529.6 XP_541529.2 1 - 2 66 65

View Full Product Details