Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC26
Entrez Gene ID
481679
Gene Name
cell division cycle 26
Gene Aliases
-
Chromosome Location
Chr.11: 67632887 - 67641291 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_538801.6 XP_538801.2 3 - 4 404 68

View Full Product Details