Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX2
Entrez Gene ID
487007
Gene Name
peroxisomal biogenesis factor 2
Gene Aliases
PXMP3
Chromosome Location
Chr.29: 24943563 - 24960834 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_544136.6 XP_544136.2 1 - 2 250 119

View Full Product Details