The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080897.1 | NP_001074366.1 | 3 - 4 | 192 | 62 |
XM_005636022.3 | XP_005636079.1 | 4 - 5 | 835 | 62 | |
XM_005636023.3 | XP_005636080.1 | 3 - 4 | 718 | 62 | |
GenBank mRNA | EF195231.1 | - | 3 - 4 | 192 | 62 |