Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MMP7
Entrez Gene ID
489432
Gene Name
matrix metallopeptidase 7
Gene Aliases
-
Chromosome Location
Chr.5: 29187902 - 29196157 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242726.1 NP_001229655.1 2 - 3 338 80
GenBank mRNA DN873521.1 - 1 - 2 236 80
JF412032.1 - 1 - 2 241 80

View Full Product Details