Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CALN1
Entrez Gene ID
607123
Gene Name
calneuron 1
Gene Aliases
-
Chromosome Location
Chr.6: 1545151 - 2067927 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003434742.4 XP_003434790.1 - 703 70
XM_005620933.3 XP_005620990.1 - 900 70
XM_005620935.3 XP_005620992.1 - 645 70
XM_014114152.2 XP_013969627.1 - 926 70
XM_014114153.2 XP_013969628.1 - 649 70
XM_022419528.1 XP_022275236.1 - 683 70

View Full Product Details