Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB26
Entrez Gene ID
491342
Gene Name
zinc finger and BTB domain containing 26
Gene Aliases
-
Chromosome Location
Chr.9: 59860499 - 59874096 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014117081.2 XP_013972556.1 1 - 2 122 86

View Full Product Details