Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYL6B
Entrez Gene ID
606992
Gene Name
myosin light chain 6B
Gene Aliases
-
Chromosome Location
Chr.10: 472415 - 476738 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014117152.2 XP_013972627.1 - 405 83
XM_843490.5 XP_848583.1 - 372 83

View Full Product Details