Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRMT61A
Entrez Gene ID
490871
Gene Name
tRNA methyltransferase 61A
Gene Aliases
-
Chromosome Location
Chr.8: 71294129 - 71300679 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_547993.5 XP_547993.1 2 - 3 881 55

View Full Product Details