Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRSS8
Entrez Gene ID
102157328
Gene Name
serine protease 8
Gene Aliases
-
Chromosome Location
Chr.6: 17110251 - 17114564 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005621242.3 XP_005621299.1 - 544 61

View Full Product Details