Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXB5
Entrez Gene ID
100686000
Gene Name
homeobox B5
Gene Aliases
-
Chromosome Location
Chr.9: 24853759 - 24856478 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003435257.4 XP_003435305.1 - 672 66

View Full Product Details