Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYT14
Entrez Gene ID
480016
Gene Name
synaptotagmin 14
Gene Aliases
-
Chromosome Location
Chr.7: 8612458 - 8795812 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022420867.1 XP_022276575.1 - 560 99

View Full Product Details