Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MED21
Entrez Gene ID
477664
Gene Name
mediator complex subunit 21
Gene Aliases
SURB7
Chromosome Location
Chr.27: 20611263 - 20619215 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197311.1 NP_001184240.1 3 - 4 277 109
XM_005636700.3 XP_005636757.1 3 - 4 350 109
GenBank mRNA GU811151.1 - 3 - 4 277 109

View Full Product Details