Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC15
Entrez Gene ID
609963
Gene Name
thioredoxin domain containing 15
Gene Aliases
-
Chromosome Location
Chr.11: 22943965 - 22958851 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_847320.4 XP_852413.1 4 - 5 936 80

View Full Product Details