Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSRA
Entrez Gene ID
608103
Gene Name
methionine sulfoxide reductase A
Gene Aliases
-
Chromosome Location
Chr.25: 27608987 - 28043461 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014107567.2 XP_013963042.1 3 - 4 727 84
XM_014107568.1 XP_013963043.1 3 - 4 219 84
XM_014107569.2 XP_013963044.1 3 - 4 231 84
XM_014107570.2 XP_013963045.1 3 - 4 632 84
XM_014107571.2 XP_013963046.1 3 - 4 367 84
XM_014107572.2 XP_013963047.1 4 - 5 700 84
XM_022409753.1 XP_022265461.1 2 - 3 304 84

View Full Product Details