The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_022405409.1 | XP_022261117.1 | 5 - 6 | 1246 | 52 |
XM_022405410.1 | XP_022261118.1 | 4 - 5 | 1111 | 52 | |
XM_540610.6 | XP_540610.2 | 4 - 5 | 1075 | 52 |