Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRPF38B
Entrez Gene ID
613001
Gene Name
pre-mRNA processing factor 38B
Gene Aliases
-
Chromosome Location
Chr.6: 43044322 - 43052622 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022420382.1 XP_022276090.1 6 - 7 843 94
XM_849600.5 XP_854693.1 4 - 5 639 94

View Full Product Details