Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMED6
Entrez Gene ID
610951
Gene Name
transmembrane p24 trafficking protein 6
Gene Aliases
-
Chromosome Location
Chr.5: 80305804 - 80313307 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_848544.4 XP_853637.1 1 - 2 1629 97

View Full Product Details