Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPH3
Entrez Gene ID
610533
Gene Name
diphthamide biosynthesis 3
Gene Aliases
-
Chromosome Location
Chr.23: 26522367 - 26529808 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_848056.5 XP_853149.1 2 - 3 258 73

View Full Product Details