Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABARAPL1
Entrez Gene ID
612919
Gene Name
GABA type A receptor associated protein like 1
Gene Aliases
-
Chromosome Location
Chr.27: 35783203 - 35791576 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005637161.2 XP_005637218.1 3 - 4 574 85

View Full Product Details