Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPAG8
Entrez Gene ID
102152667
Gene Name
sperm associated antigen 8
Gene Aliases
-
Chromosome Location
Chr.11: 52339338 - 52341642 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005626788.3 XP_005626845.1 4 - 5 1084 67

View Full Product Details