Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COX6A2
Entrez Gene ID
479780
Gene Name
cytochrome c oxidase subunit 6A2
Gene Aliases
COXVIAH
Chromosome Location
Chr.6: 16807094 - 16807763 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_536907.6 XP_536907.1 - 273 62

View Full Product Details