Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIVA1
Entrez Gene ID
612693
Gene Name
SIVA1 apoptosis inducing factor
Gene Aliases
-
Chromosome Location
Chr.8: 72309400 - 72313726 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_850420.5 XP_855513.1 3 - 4 519 129

View Full Product Details