Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RALB
Entrez Gene ID
483881
Gene Name
RAS like proto-oncogene B
Gene Aliases
-
Chromosome Location
Chr.19: 29820006 - 29868345 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005631904.3 XP_005631961.1 2 - 3 190 127
XM_541001.5 XP_541001.2 2 - 3 311 127

View Full Product Details