Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLX
Entrez Gene ID
607668
Gene Name
MAX dimerization protein MLX
Gene Aliases
TF4
Chromosome Location
Chr.9: 20391762 - 20395792 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003435229.4 XP_003435277.3 - 163 71
XM_005624449.3 XP_005624506.2 - 329 71

View Full Product Details