Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POP4
Entrez Gene ID
612732
Gene Name
POP4 homolog, ribonuclease P/MRP subunit
Gene Aliases
-
Chromosome Location
Chr.1: 121809448 - 121815811 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005616903.2 XP_005616960.2 2 - 3 143 81

View Full Product Details