Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UQCRH
Entrez Gene ID
608530
Gene Name
ubiquinol-cytochrome c reductase hinge protein
Gene Aliases
-
Chromosome Location
Chr.15: 14082954 - 14092690 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014119566.2 XP_013975041.1 1 - 2 129 116

View Full Product Details