Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTIF3
Entrez Gene ID
477328
Gene Name
mitochondrial translational initiation factor 3
Gene Aliases
-
Chromosome Location
Chr.25: 12157301 - 12171109 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_844574.5 XP_849667.1 - 841 126

View Full Product Details